Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36570

Sigma-Aldrich

α,α′-Dichloro-o-xylene

purum, ≥98.0% (GC), white to light beige

Synonym(s):

1,2-Bis(chloromethyl)benzene, o-Xylylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2Cl)2
CAS Number:
Molecular Weight:
175.06
Beilstein/REAXYS Number:
2043675
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (GC)

form

solid

color

white to light beige

bp

239-241 °C (lit.)

mp

51-55 °C (lit.)
51-55 °C

SMILES string

ClCc1ccccc1CCl

InChI

1S/C8H8Cl2/c9-5-7-3-1-2-4-8(7)6-10/h1-4H,5-6H2

InChI key

FMGGHNGKHRCJLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical form

solidified mass or chunks

replaced by

Product No.
Description
Pricing

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

257.0 °F - closed cup

flash_point_c

125 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service