Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35966

Supelco

2-Nitrophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
Beilstein/REAXYS Number:
775403
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor pressure

1 mmHg ( 49.3 °C)

product line

PESTANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1ccccc1[N+]([O-])=O

InChI

1S/C6H5NO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H

InChI key

IQUPABOKLQSFBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Guan et al.
Journal of separation science, 35(4), 490-497 (2012-01-28)
The novel surface molecularly imprinted polymer (MIP) with 2-nitrophenol (2-NP) as the template has been prepared and used as the adsorbent for the solid-phase extraction (SPE). The selectivity of the polymer was checked toward several selected nitrophenols (NPs) such as
Qing Wang et al.
Journal of separation science, 35(4), 589-595 (2012-03-06)
Coupling of long-chain ionic liquid (LCIL)-based sweeping and micelle to solvent stacking (MSS) in CZE for anionic compounds was proposed. N-Cetyl-N-methylpyrrolidinium bromide (C16MPYBr) was used as a novel cationic surfactant. The capillary column was conditioned with poly(1-vinyl-3-butylimidazolium) bromide, a kind
Zhenke Zhu et al.
Chemosphere, 93(1), 29-34 (2013-06-26)
Dissolved organic matter (DOM) is widespread in aquatic and terrestrial environments. Iron is the most abundant transition metal in the Earth's crust. The biogeochemistry of iron and the strength of Fe(II) as a reducing agent while adsorbed on minerals are
Mabel Catalán et al.
Bioelectrochemistry (Amsterdam, Netherlands), 79(2), 162-167 (2010-03-23)
In this study the interaction between new benzimidazole molecules, 2-(2-nitrophenyl)-1H-benzimidazole (NB) and N-benzoyl-2-(2-nitrophenyl)-benzimidazole (BNB), with dsDNA and ssDNA was assessed at pH 7.4. Using differential pulse voltammetry at glassy carbon electrode, both molecules were electrochemically reduced due to the presence
Ozgür Aktaş et al.
Journal of hazardous materials, 177(1-3), 956-961 (2010-01-20)
The extent of cometabolic bioregeneration of activated carbons loaded with 2-nitrophenol was investigated in lab-scale batch activated sludge reactors. Bioregeneration was quantified by measuring the deterioration in adsorption capacity of a fresh activated carbon after a pre-loading and a subsequent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service