Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32716

Supelco

Codlemone

PESTANAL®, analytical standard

Synonym(s):

trans-8, trans-10-Dodecadienol, (E,E)-8,10-DDDOL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCH2(CH2)5CH2OH
CAS Number:
Molecular Weight:
182.30
Beilstein/REAXYS Number:
2325636
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C=C\C=C\CCCCCCCO

InChI

1S/C12H22O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-5,13H,6-12H2,1H3/b3-2+,5-4+

InChI key

CSWBSLXBXRFNST-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Codlemone is a sex attractant of Laspeyresia pomonella (L.). It is used as an insecticide for controlling fruit moth in orchards.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Codling Moth: Use of [TM="Codlemone"] -Baited Traps and Visual Detection of Entries to Determine Need of Sprays.
MadsenFH and Vankenti MJ
Environmental Entomology, 2(4), 677-680 (1973)
Arthur J Stipanovic et al.
Journal of agricultural and food chemistry, 52(8), 2301-2308 (2004-04-15)
The potential utility of micrometer-sized particles as controlled-release devices for the volatilization of insect pheromones for mating disruption applications is evaluated in this study for two pheromone/model compound systems (codlemone/1-dodecanol and disparlure/1,2-epoxyoctadecane). To expedite the measurement of release rates from
L L Stelinski et al.
Journal of chemical ecology, 32(7), 1527-1538 (2006-05-24)
The effect of brief pheromone exposures on responses of codling moth (Cydia pomonella L.) males was tested by flight-tunnel and electroantennogram (EAG) studies. Males were preexposed to pheromone for up to 3 min as they sat in release cages or
D E Fernández et al.
Journal of insect science (Online), 10, 139-139 (2010-10-05)
Abstract Studies were conducted in apple, Malus domestica Borkhausen and pear, Pyrus communis L. (Rosales: Rosaceae), orchards to evaluate the attractiveness of grey halobutyl septa loaded with 1 (L2) and 10 (Mega) mg of codlemone, 8E, 10E-dodecadien-1-ol, 3 mg of
Elizabeth Tomaszewska et al.
Journal of agricultural and food chemistry, 53(7), 2399-2405 (2005-03-31)
Pome fruit growers and crop consultants have expressed concerns about the seasonal release performance of commercial codling moth mating disruption dispenser products. Because of these concerns, we developed a laboratory flow-through volatile collection system (VCS) for measuring the volatile release

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service