Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32352

Supelco

Chromafenozide

PESTANAL®, analytical standard

Synonym(s):

2′-tert-Butyl-5-methyl-2′-(3,5-xyloyl)chromane-6-carbohydrazide, 3,4-Dihydro-5-methyl-2H-1-benzopyran-6-carboxylic acid 2-(3,5-dimethylbenzoyl)-2-(1,1-dimethylethyl)hydrazide, N-(5-Methylchroman-6-carbonyl)-N′-tert-butyl-N′-(3,5-dimethylbenzoyl)hydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30N2O3
CAS Number:
Molecular Weight:
394.51
Beilstein/REAXYS Number:
10113703
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(NN(C(C)(C)C)C(C1=CC(C)=CC(C)=C1)=O)C2=CC=C3C(CCCO3)=C2C

InChI

1S/C24H30N2O3/c1-15-12-16(2)14-18(13-15)23(28)26(24(4,5)6)25-22(27)20-9-10-21-19(17(20)3)8-7-11-29-21/h9-10,12-14H,7-8,11H2,1-6H3,(H,25,27)

InChI key

HPNSNYBUADCFDR-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ehab Abdelraheem et al.
Environmental monitoring and assessment, 189(4), 180-180 (2017-03-28)
The objective of this study is to perform a safety assessment of chromafenozide residue level on tomato at the Egyptian national level. An open field decline study of chromafenozide on tomato was performed. The theoretical maximum daily intake (TMDI) of
Jinhua Gan et al.
Ecotoxicology and environmental safety, 210, 111853-111853 (2021-01-11)
A multi-residue analysis of six diacylhydrazine insecticides in water, sediment, and aquatic products was established by liquid chromatography triple quadrupole tandem mass spectrometry (LC-MS/MS). The water sample was extracted with acetonitrile by low-temperature enrichment liquid-liquid extraction technology. The sediment and
Sayoko Ito-Harashima et al.
FEBS open bio, 7(7), 995-1008 (2017-07-07)
Synthetic nonsteroidal ecdysone agonists, a class of insect growth regulators (IGRs), target the ecdysone receptor (EcR), which forms a heterodimer with ultraspiracle (USP) to transactivate ecdysone response genes. These compounds have high binding affinities to the EcR-USP complexes of certain
Naoki Okamoto et al.
Current biology : CB, 30(2), 359-366 (2020-01-14)
Steroid hormones control various aspects of brain development and behavior in metazoans, but how they enter the central nervous system (CNS) through the blood-brain barrier (BBB) remains poorly understood. It is generally believed that steroid hormones freely diffuse through the
Shiming Song et al.
Journal of chromatography. A, 1557, 51-61 (2018-05-08)
In this study, we developed an anion exchanger-disposable pipette extraction (DPX) method to detect the residual concentrations of eight neonicotinoid insecticides (dinotefuran, acetamiprid, clothianidin, thiacloprid, imidachloprid, imidaclothiz, nitenpyram, and thiamethoxam) and eight insect growth regulators (IGRs; triflumuron, cyromazine, buprofezin, methoxyfenozide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service