Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31570

Supelco

2-Chloro-4-nitrotoluene

analytical standard

Synonym(s):

2-Chloro-1-methyl-4-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)Cl
CAS Number:
Molecular Weight:
171.58
Beilstein/REAXYS Number:
1817924
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

60-64 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1ccc(cc1Cl)[N+]([O-])=O

InChI

1S/C7H6ClNO2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,1H3

InChI key

LLYXJBROWQDVMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

212.0 °F - closed cup

flash_point_c

100 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gundula Wolf et al.
Journal of biotechnology, 128(4), 801-812 (2007-02-13)
An improved method for deconvoluting complex spectral maps from bidimensional fluorescence monitoring is presented, relying on a combination of principal component analysis (PCA) and feedforward artificial neural networks (ANN). With the aim of reducing ANN complexity, spectral maps are first
[Method of determining p-nitrotoluene, 2-chloro-4-nitrotoluene and 3-chloro-4-methylaniline in the air].
I I Pilenkova et al.
Gigiena truda i professional'nye zabolevaniia, (1)(1), 43-44 (1980-01-01)
G Wolf et al.
Biotechnology and bioengineering, 72(3), 297-306 (2001-01-03)
The use of two-dimensional scanning fluorometry as an on-line, noninvasive, in situ bioreactor monitoring technique is extended to complex bioprocesses using mixed cultures, with particular attention to biofilm systems. Using the example of spectra subtraction, it is demonstrated that established

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service