Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26585

Sigma-Aldrich

5-Chlorovaleroyl chloride

purum, ≥97.0% (T)

Synonym(s):

5-Chloropentanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)4COCl
CAS Number:
Molecular Weight:
155.02
Beilstein/REAXYS Number:
1745182
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (T)

refractive index

n20/D 1.464 (lit.)
n20/D 1.464

bp

38-39 °C/0.15 mmHg (lit.)

density

1.206 g/mL at 20 °C
1.2 g/mL at 25 °C (lit.)

SMILES string

ClCCCCC(Cl)=O

InChI

1S/C5H8Cl2O/c6-4-2-1-3-5(7)8/h1-4H2

InChI key

SVNNWKWHLOJLOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service