Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25860

Sigma-Aldrich

4-Chlorophenol

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OH
CAS Number:
Molecular Weight:
128.56
Beilstein/REAXYS Number:
507004
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor pressure

1 mmHg ( 49.8 °C)

grade

purum

assay

≥98.0% (GC)

bp

220 °C (lit.)

mp

40-45 °C
42-45 °C

density

1.306 g/mL at 25 °C (lit.)

SMILES string

Oc1ccc(Cl)cc1

InChI

1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H

InChI key

WXNZTHHGJRFXKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick W Causey et al.
Journal of medicinal chemistry, 51(9), 2833-2844 (2008-04-17)
A series of mono and diaryl rhenium(I)-carborane derivatives were prepared using microwave heating and screened for their affinity for two isoforms of the estrogen receptor (ER). The rhenacarborane derivative [(RR'C 2B9H9)Re(CO)3](-) (R = p-PhOH, R' = H), which was generated
Giorgio Ottaviani et al.
Journal of medicinal chemistry, 49(13), 3948-3954 (2006-06-23)
This work was devoted to the search for new artificial membranes allowing a rapid evaluation of passive human skin permeation of compounds with a parallel artificial membrane permeability assay (PAMPA). Effective permeability coefficients (Pe) determined for a set of compounds
Giorgio Ottaviani et al.
Journal of medicinal chemistry, 50(4), 742-748 (2007-02-16)
The development of in silico and in vitro tools to estimate or predict the passive human skin permeation and distribution of new chemical entities, useful in dermal drug delivery, in absorption studies of toxic compounds, and in the cosmetics industry
Cynthia D Selassie et al.
Journal of medicinal chemistry, 48(23), 7234-7242 (2005-11-11)
In this comprehensive study on the caspase-mediated apoptosis-inducing effect of 51 substituted phenols in a murine leukemia cell line (L1210), we determined the concentrations needed to induce caspase activity by 50% (I50) and utilized these data to develop the following
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service