Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23730

Sigma-Aldrich

2-Chlorobenzoyl chloride

purum, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COCl
CAS Number:
Molecular Weight:
175.01
Beilstein/REAXYS Number:
386435
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (HPLC)

refractive index

n20/D 1.572 (lit.)
n20/D 1.572

bp

238 °C (lit.)

mp

−4-−3 °C (lit.)

density

1.382 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1ccccc1Cl

InChI

1S/C7H4Cl2O/c8-6-4-2-1-3-5(6)7(9)10/h1-4H

InChI key

ONIKNECPXCLUHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 103918-ALDRICH | 2-Chlorobenzoyl chloride 95%

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

255.2 °F - closed cup - DIN 51758

flash_point_c

124 °C - closed cup - DIN 51758

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service