14610
Methyl acetoacetate
analytical standard
Synonym(s):
3-Oxobutanoic acid methyl ester, Acetoacetic acid methyl ester
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥99.5% (GC)
autoignition temp.
536 °F
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.419 (lit.)
n20/D 1.419
bp
169-170 °C/70 mmHg (lit.)
mp
−80 °C (lit.)
density
1.076 g/mL at 25 °C (lit.)
application(s)
environmental
format
neat
SMILES string
COC(=O)CC(C)=O
InChI
1S/C5H8O3/c1-4(6)3-5(7)8-2/h3H2,1-2H3
InChI key
WRQNANDWMGAFTP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Recommended products
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
158.0 °F - closed cup
flash_point_c
70 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service