13738
(S)-(−)-3-Benzyloxy-1,2-propanediol
puriss., ≥99.0% (sum of enantiomers, GC)
Synonym(s):
(S)-Glycerol 1-benzyl ether, 1-Benzyl-sn-glycerol
About This Item
grade
puriss.
assay
≥99.0% (sum of enantiomers, GC)
form
solid
optical activity
[α]20/D −5.5±0.5°, c = 20% in chloroform
bp
261 °C (lit.)
mp
24-26 °C (lit.)
storage temp.
2-8°C
SMILES string
OC[C@H](O)COCc1ccccc1
InChI
1S/C10H14O3/c11-6-10(12)8-13-7-9-4-2-1-3-5-9/h1-5,10-12H,6-8H2/t10-/m0/s1
InChI key
LWCIBYRXSHRIAP-JTQLQIEISA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service