Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09895

Sigma-Aldrich

Fmoc-β-phenyl-Phe-OH

≥98.0% (HPLC)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-β-phenyl-L-phenylalanine, Fmoc-3,3-diphenyl-L-alanine, Fmoc-β,β-diphenyl-Ala-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H25NO4
CAS Number:
Molecular Weight:
463.52
Beilstein/REAXYS Number:
8873562
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Quality Level

assay

≥98.0% (HPLC)

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white to almost white

mp

125-129 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(c4ccccc4)c5ccccc5

InChI

1S/C30H25NO4/c32-29(33)28(27(20-11-3-1-4-12-20)21-13-5-2-6-14-21)31-30(34)35-19-26-24-17-9-7-15-22(24)23-16-8-10-18-25(23)26/h1-18,26-28H,19H2,(H,31,34)(H,32,33)/t28-/m0/s1

InChI key

PENQOTJCVODUQU-NDEPHWFRSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service