07296
Tris(2-chloroethyl) phosphate
analytical standard
Synonym(s):
Phosphoric acid tris(2-chloroethyl) ester
About This Item
Recommended Products
grade
analytical standard
Quality Level
vapor pressure
<10 mmHg ( 25 °C)
assay
≥95.0% (GC)
shelf life
limited shelf life, expiry date on the label
refractive index
n20/D 1.472 (lit.)
bp
192 °C/10 mmHg (lit.)
density
1.39 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
neat
SMILES string
ClCCOP(=O)(OCCCl)OCCCl
InChI
1S/C6H12Cl3O4P/c7-1-4-11-14(10,12-5-2-8)13-6-3-9/h1-6H2
InChI key
HQUQLFOMPYWACS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
449.6 °F - closed cup
flash_point_c
232 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service