Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03710

Sigma-Aldrich

Ethylenediaminetetraacetic acid trisodium salt dihydrate

≥98.0% (KT)

Synonym(s):

Trisodium ethylenediaminetetraacetate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N2Na3O8 · 2H2O
CAS Number:
Molecular Weight:
394.22
Beilstein/REAXYS Number:
3858591
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Product 03710 is not currently sold in your country. Contact Technical Service

assay

≥98.0% (KT)

SMILES string

O.O.[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.3Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;2*1H2/q;3*+1;;/p-3

Inchi Key

OABSPZRQQUFHRY-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2 Inhalation

target_organs

Respiratory Tract

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service