Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00803

Sigma-Aldrich

Acetoxyacetone

technical, ≥98.0% (GC)

Synonym(s):

Acetonyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2OCOCH3
CAS Number:
Molecular Weight:
116.12
Beilstein/REAXYS Number:
1749857
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

assay

≥98.0% (GC)

refractive index

n20/D 1.415

bp

174-176 °C (lit.)

density

1.075 g/mL at 20 °C (lit.)

SMILES string

CC(=O)COC(C)=O

InChI

1S/C5H8O3/c1-4(6)3-8-5(2)7/h3H2,1-2H3

InChI key

DBERHVIZRVGDFO-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service