Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00549

Supelco

trans-2-Dodecen-1-ol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O
CAS Number:
Molecular Weight:
184.32
Beilstein/REAXYS Number:
1758331
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

assay

≥95.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.453

density

0.846 g/mL at 20 °C (lit.)

application(s)

environmental

SMILES string

CCCCCCCCC\C=C\CO


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service