Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00351

Sigma-Aldrich

Fmoc-β-(2-furyl)-Ala-OH

≥98.0% (HPLC)

Synonym(s):

(S)-2-(Fmoc-amino)-3-(2-furyl)propionicacid, Fmoc-3-(2-furyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19NO5
CAS Number:
Molecular Weight:
377.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Quality Level

assay

≥98.0% (HPLC)

form

powder with small lumps

color

white to faintly beige

mp

136-141 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccco1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C22H19NO5/c24-21(25)20(12-14-6-5-11-27-14)23-22(26)28-13-19-17-9-3-1-7-15(17)16-8-2-4-10-18(16)19/h1-11,19-20H,12-13H2,(H,23,26)(H,24,25)/t20-/m0/s1

InChI key

AJXDCHXGNUFBRC-FQEVSTJZSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service