Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX0692

Supelco

Sodium Perchlorate

anhydrous, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Sodium Perchlorate, Perchloric Acid, Sodium Salt, Perchloric Acid Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClNaO4
CAS Number:
Molecular Weight:
122.44
MDL number:
UNSPSC Code:
12352300
EC Index Number:
231-511-9
assay:
98.0-102.0% (silver nitrate titration)
grade:
ACS reagent
form:
crystals

grade

ACS reagent

Quality Level

agency

meets USP/NF

assay

98.0-102.0% (silver nitrate titration)

form

crystals

impurities

≤0.005% Insoluble matter

color

white

pH

6.0-8.0 (25 °C, 5% in solution)

mp

471.8 °C

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.020%
Fe: ≤5 ppm
K: ≤0.050%
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

InChI

1S/ClHO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

BAZAXWOYCMUHIX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

anhydrous, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (argentometric): 98.0-102.0%
Calcium (Ca): 0.02% max
Chloride (Cl): 0.003% max
Color: White
Form: Crystals
Heavy Metals (by ICP-OES): 5ppm max
Insoluble matter: 0.005% max
Iron (Fe): 5ppm max
Potassium (K): 0.05% max
Sulfate (SO4): 0.002% max
pH of a 5% solution at 25°C: 6.0-8.0

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1 - STOT RE 2

target_organs

Thyroid

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service