Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R03266

Sigma-Aldrich

Lactophenol Solution

in accordance with BSC

Synonym(s):

Parasitology Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130

agency

in accordance with BSC

Quality Level

color

colorless

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

General description

Lactophenol Solution
R03266-74 500ml Glass Bottle
Used in parasitology

Application

Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used in parasitology.
Used in parasitology

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

target_organs

Nervous system,Kidney,Liver,Skin

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service