Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC260

Sigma-Aldrich

Anti-TRAF1 Antibody, clone 1F3

clone 1F3, from rat

Synonym(s):

TNF receptor-associated factor 1, TNF receptor-associated factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$423.00

$423.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$423.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$423.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F3, monoclonal

species reactivity

monkey, human

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRAF1(7185)

Related Categories

General description

TNF receptor-associated factor 1 (UniProt: Q13077; also known as Epstein-Barr virus-induced protein 6, TRAF1) is encoded by the TRAF1 (also known as EB16) gene (Gene ID: 7185) in human. TRAF1 is homodimeric pro-survival adapter molecule that regulates the activation of NF-kB and JNK. It can also exist as a heterotrimer with TRAF2. It plays a role in the regulation of cell survival and apoptosis. The heterotrimer form is part of a E3 ubiquitin-protein ligase complex that promotes ubiquitination of target proteins, such as MAP3K14. The TRAF1/TRAF2 complex recruits the antiapoptotic E3 protein-ubiquitin ligases BIRC2 and BIRC3 to TNFRSF1B/TNFR2. The C-terminal region of TRAF1 contains the TRAF domain (aa 266-412) that binds to receptor cytoplasmic domains. It also has a coiled coil domain (aa 182-264) that mediates its homo- and heterotrimerization. TRAF1 is also reported to essential for memory T cell survival. TRAF1 protein level is significantly reduced in HIV-specific CD8 T cells with progression of infection. Its levels can increase following treatment with IL-7. Hence, TRAF1 levels could serve as a useful parameter in monitoring effectiveness of anti-HIV therapies. (Ref.: Wang, C., et al. (2018). J. Immunol. 200(2); 558-564).

Immunogen

GST-tagged recombinant protein corresponding to human TRAF1.

Application

Anti-TRAF1, clone 1F3 detects levels of TRAF1 proteins & has been published & validated for use in WB, IHC & Flow.
Immunohistochemistry Analysis: A representative lot was used by an an independent laboratory to detect TRAF1 in certain cell lines (Siegler, G., et al. (2003). J Clin Pathol: Mol Pathol. 56:156-161.).
Flow Cytometry Analysis: A representative lot was used by an an independent laboratory to detect TRAF1 in PBMCs and in certain T cells from HIV-infected individuals (Wang, C., et al. (2011). JEM.1-15.).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in B95-8 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected TRAF1 in 10 µg of B95-8cell lysate.

Target description

~46 kDa observed. Uniprot describes an isoform produced by alternative splicing at ~33 kDa

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
B95-8cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Wang et al.
Cell genomics, 3(11), 100420-100420 (2023-11-29)
TRAF1/C5 was among the first loci shown to confer risk for inflammatory arthritis in the absence of an associated coding variant, but its genetic mechanism remains undefined. Using Immunochip data from 3,939 patients with juvenile idiopathic arthritis (JIA) and 14,412

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service