Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB327F

Sigma-Aldrich

Anti-Alzheimer Precursor Protein A4 (NT)-FITC Antibody, clone 22C11

clone 22C11, Milli-Mark®, from mouse

Synonym(s):

APP

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$365.00

$365.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$365.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$365.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

22C11, monoclonal

species reactivity

canine

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

General description

Deposits of amyloid protein in senile plaques near nerve processes are found in the brains of aged humand and cases of Alzheimer′s Disease. The principle component of this extracellular amyloid is beta A4, a 4 kDa peptide derived from a larger amyloid precursor protein (APP), which is widely expressed in the brain and body. The creation, transport and function of these proteins is currently an active area of research.

Specificity

Antibody recognizes Alzheimer Precursor Protein

Immunogen

Purified recombinant Alzheimer precursor A4 (pre A4695) fusion protein.

Application

Anti-Alzheimer Precursor Protein A4 (N-terminus)-FITC Antibody, clone 22C11 detects level of Alzheimer Precursor Protein A4 (N-terminus) & has been published & validated for use in FC.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by flow cytometry using B35 cells

Target description

110/120/130 kDa Calculated

Physical form

Protein A purified
Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light for up to 6 months from date of receipt.

Analysis Note

Control
B35 cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service