Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB140P

Sigma-Aldrich

Milli-Mark® Anti-phospho-Bcl-2 (Ser70) -PE Antibody, clone 69-10C-2-10C-18

clone 69-10C-2-10C-18, Milli-Mark®, from rabbit

Synonym(s):

B-cell CLL/lymphoma 2

B-cell lymphoma protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$301.00

$301.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$301.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$301.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

69-10C-2-10C-18, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer70)

Gene Information

human ... BCL2(596)

Related Categories

General description

The Bcl name is derived from B-cell chronic lymphocytic leukemia, and was given to genes involved in that disease. Members of the Bcl-2 family share homologous domains (BH) through which they are able to homo- and hetero-dimerize. Bcl-2 plays an important role in cytochrome c release from the mitochondria; it also binds Apaf1, inhibiting Apaf-1 association with caspase-9. Bcl-2 overexpression in indicated in a number of human cancers, including melanoma, breast, prostate, and lung, and it correlates with resistance to cancer treatment (presumably due to decreased apoptosis). Bcl-2 has also been implicated in schizophrenia and autoimmune disorders. Bcl-2 phosphorylated at Ser-70 plays a critial role in the translocation of Bax from the cytosol to the mitochondria, and this may regulate NO-induced neuronal apoptosis. The molecular mechanism of Bcl-2 phosphorylation and its relationship to Bax is largely unknown.

Specificity

This antibody recognizes phosphorylated Bcl-2 at Ser70.

Immunogen

Epitope: Ser70
KLH-conjugated linear peptide corresponding to human Bcl-2 phosphorylated at Ser70.

Application

Detect phospho-Bcl-2 (Ser70) using this Milli-Mark Anti-phospho-Bcl-2 Antibody, clone 69-10C-2-10C-18 validated for use in FC.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition

Quality

Evaluated by flow cytometry using Jurkat cells.

Target description

26 kDa Observed

Physical form

Protein A purified
Purified rabbit monoclonal IgG conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Jurkat cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service