Key Documents
TANKVNT21
Milli-Q
Vent Filter
pore size 0.22 μm, Advanced protection against airborne contaminants for storage tanks
About This Item
Product TANKVNT21 is not currently sold in your country. Contact Technical Service
Quality Level
usage
(for type 2 water)
packaging
pkg of 2 units
manufacturer/tradename
Milli-Q®
H
17 cm (6.7 in.)
pore size
0.22 μm pore size
compatibility
for use with SDS
for use with tanks
General description
  • The vent filter protects highly sensitive stored pure water from possible contaminants in the environment. These include carbon dioxide, particles, micro-organisms and volatile organic compounds that mainly come from the laboratory atmosphere.
  • Vent filters contain a combination of purification media carefully selected to remove airborne contaminants that could degrade purified water quality during storage.
  • Vent filter without soda lime.
  • The vent filter cartridges are constructed from polypropylene, polyethersulfone and styrene-acrylonitrile resin.
Application
The vent filter provides advanced protection against airborne contaminants.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany