Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABT404

Sigma-Aldrich

Anti-hEF1/NEDD9, clone 2G9 Antibody

clone 2G9, 1 mg/mL, from mouse

Synonym(s):

Enhancer of filamentation 1, hEF1, CRK-associated substrate-related protein, CAS-L, CasL, Cas scaffolding protein family member 2, Neural precursor cell expressed developmentally down-regulated protein 9, NEDD-9, Renal carcinoma antigen NY-REN-12, p105,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2G9, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NEDD9(4739)

General description

Enhancer of filamentation 1, also known as CRK-associated substrate-related protein (CAS-L) or Cas scaffolding protein family member 2 or Neural precursor cell expressed developmentally down-regulated protein 9 (NEDD-9), or Renal carcinoma antigen NY-REN-12 p105, and encoded by the gene NEDD9 or CASL or CASS2, is a docking protein that plays a central role in coordinating tyrosine kinase mediated signaling for cell adhesion. NEDD9 controls the signals between focal adhesions at the cell periphery and the mitotic spindle in response to growth factors and signals. NEDD9 interacts with integrins and various other cellular proteins including BCAR3 and SH2D3C. It is localized to the cytoplasm in most cells but location can vary depending upon the cell type and function. NEDD9 is also found in cellular projections and lamellipodium and is found in the nucleus depending upon the cellular microenvironment. NEDD9 has multiple isoforms and active proteolytic fragments ranging from 55 kDa to 115 kDa. Isoforms p105 and p115 are predominantly cytoplasmic and associate with focal adhesions while p55 associates with the mitotic spindle. NEDD9 is highly expressed in most cells and tissues and is typically induced upon cell growth and down regulated in adult tissues such as brain. NEDD9 is processed during the cell cycle into multiple proteolytic fragments that appear to have important functions at specific times during the cell cycle process. In cancer NEDD9 over expression can promote tumor growth and NEDD9 acts directly to stabilize Aurora A kinase which is often over expressed in many cancers. Destabilization of Aurora A via the inhibition or destruction of NEDD9 allows for tumor reduction, thus NEDD9 expression is being examined as a potential cancer marker.

Immunogen

Recombinant protein corresponding to human hEF1/NEDD9.

Application

Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-hEF1/NEDD9, clone 2G9 Antibody is validated for use in Western Blotting for the detection of hEF1/NEDD9.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected hEF1/NEDD9 in 10 µg of SH-SY5Y cell lysate.

Quality

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected hEF1/NEDD9 in 10 µg of MCF-7 cell lysate.

Target description

~93 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service