Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC112S

Sigma-Aldrich

Sheep Anti-Human IgG Antibody, Cy5 conjugate

from sheep, CY5 conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

sheep

conjugate

CY5 conjugate

antibody form

purified antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) is part of the family of glycoprotein immunoglobulins, which stage immune responses against pathogenic organisms. IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. IgG contributes to the immune response by mobilizing innate immune effector cells and inducing and pro-inflammatory responses. Such proinflammatory responses are mediated by Fc gamma receptors which interact with sialylated Fc fragments on IgG molecules. The coupling of Cy5 to Anti-Human IgG antibody allows for the visualization of proteins by fluorescent microscopy.

Specificity

Based on ELISA analysis using the unconjugated antibody (Cat. No. AC112), this antibody exhibits strongest immunoreactivity towards whole human IgG molecule (IgG>IgM>IgG Fc>IgG Fab>IgA>IgM Fc) with minimal or no cross-reactivity toward IgG from chicken, guinea pig, hamster, mouse, rabbit, and rat species.

Immunogen

Purified human IgG.

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
All
This Sheep Anti-Human IgG, Cy5 conjugate, Cat. No. AC112S, detects human IgG and is validated for Immunocytochemistry staining of human IgG-producing B-cell hybridoma SA13.
This antibody is also available in unconjugated and Cy3-conjugated forms (Cat. Nos. AC112 and AC112C).

Quality

Evaluated by Immunocytochemistry in human B-cell hybridoma SA13.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody immunostained 4% paraformaldehyde-fixed, 0.1% Triton X-100-permeabilized human B-cell hybridoma SA13, but not HeLa cells.

Target description

~150 kDa observed.

Physical form

Ammonium sulfate.
Purified sheep polyclonal antibody Cy5 conjugate in PBS with 1.5% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service