Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABF1215-AF488

Sigma-Aldrich

Anti-SLP76 Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

Lymphocyte cytosolic protein 2, SH2 domain-containing leukocyte protein of 76 kDa, SLP-76 tyrosine phosphoprotein, SLP76

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

flow cytometry: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LCP2(3937)

General description

Lymphocyte cytosolic protein 2 (UniProt: Q13094; also known as Lymphocyte cytosolic protein 2, SH2 domain-containing leukocyte protein of 76 kDa, SLP-76 tyrosine phosphoprotein, SLP76) is encoded by the gene LCP2 (Gene ID 3937) in human. SLP-76 is a cytoplasmic adaptor phosphoprotein that contains both SH2 and SAM domains and is involved in B and T cell receptor signaling. Its amino terminus contains an acidic region that includes a PEST domain and several tyrosine residues that are phosphorylated following TCR ligation. SLP-76 also contains a central proline-rich domain and a SH2 domain at its carboxy terminal. SLP-76 can be phosphorylated on multiple tyrosine residues by the upstream kinases ZAP-70 and Lck. SLP-76 phosphorylation plays an important role in T cell-mediated IL-2 production by allowing phosphorylated Vav to bind and this complex stimulates NFAT and IL-2 gene activation after TCR engagement. SLP-76 is highly expressed in spleen, thymus, and peripheral blood leukocytes and T cells. It is expressed in B cell lines at lower levels and is not detected in fibroblast and neuroblastoma cell lines. SLP-76 is reported to play a positive role in promoting T cell development and activation as well as mast cell and platelet function. Overexpression of SLP-76 is shown to enhance IL-2 transcription after TCR signaling.

Specificity

This rabbit polyclonal antibody Alexa Fluor 488 conjugate detected SLP76 (lymphocyte cytosolic protein 2) in Jurkat cells by flow cytometry.

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of human SLP76.

Application

Flow Cytometry Analysis: 1:400 dilution from a representative lot detected SLP76 in 4% paraformaldeyde-fixed, 0.2% Triton X-100-permeabilized THP-1 human monocytic cells.

The unconjugated antibody (Cat. No. ABF1215) is also available for Western blotting application.
This rabbit polyclonal Anti-SLP76, Alexa Fluor 488 Conjugate, Cat. No. ABF1215-AF488 is validated for use in Flow Cytometry for the detection of SLP76 (lymphocyte cytosolic protein 2).

Quality

Evaluated by Flow Cytometry in Jurkat cells.

Flow Cytometry Analysis: A 1:400 diluution of this antibody detected SLP76 in 4% paraformaldeyde-fixed, 0.2% Triton X-100-permeabilized Jurkat human T lymphocytes.

Target description

60.2 kDa calculated.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service