Concentrate urine and cerebrospinal fluid to intensify proteins that indicate abnormal or pathological states prior to analysis by electrophoresis or immunoelectrophoresis
Static concentrator, requiring no accessories
Absorbent pulls solvent and salts through ultrafilter, concentrating sample
Applications
Concentration of Urine and Cerebrospinal Fluid for Enrichment of Protein Analytes
Ultrafiltration
Use the Minicon® static concentrators to concentrate urine and cerebrospinal fluid to intensify proteins that indicate abnormal or pathological states prior to analysis by electrophoresis or immunoelectrophoresis. The B15 unit has 8 cells and can hold up to 5 mL of sample. The CS15 has 10 cells and is suited for volumes up to 2.5 mL.
Features and Benefits
Minimum Final Concentrate Volume: 30 µL
Static concentrator, requiring no accessories
Absorbent pulls solvent and salts through ultrafilter, concentrating sample
Concentration factor up to 100X
Low hold-up volume
Physical form
Clear
Legal Information
MINICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.