Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45139

Sigma-Aldrich

Undecanoyl chloride

Synonym(s):

Undecanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21ClO
CAS Number:
Molecular Weight:
204.74
MDL number:
UNSPSC Code:
12352101

assay

≥99.0% (GC)

Quality Level

bp

136 °C/20 mbar

transition temp

flash point 112 °C

density

0.927 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C11H21ClO/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3

InChI key

JUKPJGZUFHCZQI-UHFFFAOYSA-N

Analysis Note

Colour (visual): colourless to almost colourless
Appearance (visual): clear
Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.925 - 0.930
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service