Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.40139

Sigma-Aldrich

Phenoxyacetamide

for synthesis

Synonym(s):

Phenoxyacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
UNSPSC Code:
12352111
EC Index Number:
210-713-0

Quality Level

assay

≥94.0% (HPLC)

mp

98-102 °C

InChI

1S/C8H9NO2/c9-8(10)6-11-7-4-2-1-3-5-7/h1-5H,6H2,(H2,9,10)

InChI key

AOPRXJXHLWYPQR-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 94.0 % (a/a)
Melting range (lower value): ≥ 98 °C
Melting range (upper value): ≤ 102 °C
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service