8.18452
DL-Menthol
for synthesis
Synonym(s):
DL-Menthol, Hexahydrothymol, 2-Isopropyl-5-methylcyclohexanol
About This Item
Recommended Products
vapor pressure
130 Pa ( 55 °C)
Quality Level
assay
≥99.0% (GC)
autoignition temp.
405 °C
potency
3180 mg/kg LD50, oral (Rat)
bp
212 °C/1013 hPa
mp
31-34 °C
transition temp
flash point 96 °C
density
0.89 g/cm3 at 20 °C
InChI
1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1
InChI key
NOOLISFMXDJSKH-KXUCPTDWSA-N
Analysis Note
Melting range (lower value): ≥ 31 °C
Melting range (upper value): ≤ 35 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
204.8 °F
flash_point_c
96 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service