Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14552

Sigma-Aldrich

Sodium hydride suspension

(60% suspension in paraffin oil) for synthesis

Synonym(s):

Sodium hydride suspension

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNa
CAS Number:
Molecular Weight:
24.00
MDL number:
UNSPSC Code:
12352306
EC Index Number:
231-587-3

reaction suitability

reagent type: reductant

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/Na

InChI key

KEAYESYHFKHZAL-UHFFFAOYSA-N

Analysis Note

Assay (by gasvolumetry): ≥ 57.0 %

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service