Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08240

Sigma-Aldrich

Tetracyanoethylene

for synthesis

Synonym(s):

Tetracyanoethylene, Ethylene tetracarbonitrile, TCNE, Percyanoethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CN)2C=C(CN)2
CAS Number:
Molecular Weight:
128.09
MDL number:
UNSPSC Code:
12352117
EC Index Number:
211-578-0

Quality Level

assay

≥98.0% (GC)

potency

5 mg/kg LD50, oral (Rat)

mp

196-199 °C

density

1.35 g/cm3 at 25 °C

InChI

1S/C6N4/c7-1-5(2-8)6(3-9)4-10

InChI key

NLDYACGHTUPAQU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service