Skip to Content
MilliporeSigma
All Photos(2)

Documents

8.02672

Sigma-Aldrich

Hexanal (stabilised)

for synthesis

Synonym(s):

Hexanal (stabilised), Hexaldehyde, Caproaldehyde, Hexylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CHO
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352114
EC Index Number:
200-624-5

vapor pressure

12 hPa ( 20 °C)

Quality Level

autoignition temp.

220 °C

potency

4890 mg/kg LD50, oral (Rat)
5500 mg/kg LD50, skin (Rabbit)

pH

4-5 (20 °C, 4.8 g/L in H2O)

bp

129-131 °C/1013 hPa

mp

-56 °C

transition temp

flash point 25 °C

solubility

6 g/L

density

0.81 g/cm3 at 20 °C

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h6H,2-5H2,1H3

InChI key

JARKCYVAAOWBJS-UHFFFAOYSA-N

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

77.0 °F - closed cup

flash_point_c

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Zamani Ghaleshahi et al.
Food research international (Ottawa, Ont.), 130, 108966-108966 (2020-03-12)
In this work, coated liposome as a novel delivery system assembled from genipin-crosslinked with whey protein isolate (WPI) and sodium alginate (SA) as the second layer on WPI-coated liposomal was developed in order to increase its bioavailability and prolong release

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service