Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09794

Supelco

n-Undecane

reference substance for gas chromatography

Synonym(s):

n-Undecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH3
CAS Number:
Molecular Weight:
156.31
MDL number:
UNSPSC Code:
12352001
EC Index Number:
214-300-6

vapor pressure

0.6 hPa ( 20 °C)

Quality Level

assay

≥99.5% (GC)

form

liquid

autoignition temp.

220 °C

expl. lim.

0.6-6.5 % (v/v)

technique(s)

gas chromatography (GC): suitable

kinematic viscosity

1.6 cSt(20 °C)

mp

-26 °C

transition temp

flash point 60 °C

density

0.74 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C11H24/c1-3-5-7-9-11-10-8-6-4-2/h3-11H2,1-2H3

InChI key

RSJKGSCJYJTIGS-UHFFFAOYSA-N

Analysis Note

Assay (GC): ≥ 99.5 %
Identity (IR-spectrum): conforms

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

140.0 °F

flash_point_c

60 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service