Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07722

Supelco

Semicarbazide hydrochloride

GR for analysis

Synonym(s):

Semicarbazide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH5N3O · xHCl
CAS Number:
Molecular Weight:
75.07 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-247-0

form

solid

potency

123 mg/kg LD50, oral (Rat)

pH

1.5 (20 °C, 100 g/L in H2O)

mp

175-177 °C (decomposition)

bulk density

740 kg/m3

InChI

1S/CH5N3O.ClH/c2-1(5)4-3;/h3H2,(H3,2,4,5);1H

Inchi Key

XHQYBDSXTDXSHY-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.5 %
Identity (IR-spectrum): passes test
In water insoluble matter: ≤ 0.02 %
Hydrazine: ≤ 0.01 %
Alcohols (GC sum): ≤ 0.01 %
Sulfated ash: ≤ 0.02 %

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service