Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02786

Supelco

Manganese(II) sulfate tetrahydrate

for analysis EMSURE®

Synonym(s):

Manganese(II) sulfate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO4S · 4H2O
CAS Number:
Molecular Weight:
223.06
UNSPSC Code:
12352302
EC Index Number:
232-089-9
assay:
≥98.5% (complexometric)
form:
solid

Quality Level

product line

EMSURE®

assay

≥98.5% (complexometric)

form

solid

potency

2150 mg/kg LD50, oral (Rat)

pH

2.5-4.0 (20 °C, 50 g/L in H2O)

mp

26-27 °C

density

2.11 g/cm3 at 20 °C

bulk density

1000‑1200 kg/m3

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.005%
Cu: ≤0.0002%
Fe: ≤0.001%
K: ≤0.002%
Mg: ≤0.01%
Na: ≤0.005%
Ni: ≤0.002%
Pb: ≤0.0002%
Zn: ≤0.002%

storage temp.

15-25°C

InChI

1S/Mn.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

SQQMAOCOWKFBNP-UHFFFAOYSA-L

Analysis Note

Assay (complexometric): ≥ 98.5 %
pH-value (5 %; water): 4.5 - 6.0
Chloride (Cl): ≤ 0.001 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.002 %
Mg (Magnesium): ≤ 0.01 %
Na (Sodium): ≤ 0.005 %
Ni (Nickel): ≤ 0.002 %
Pb (Lead): ≤ 0.0002 %
Zn (Zinc): ≤ 0.002 %
Oxidizing matter: passes test
Reducing substances: passes test

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2 Inhalation

target_organs

Brain

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Dear whom this may concern, I recently bought this product from your company. However, I was wondering what the maximum solubility of this compound is in water? Cheers, Andre

    1 answer
    1. Unfortunately, this product is not tested for solubility. This literature suggests that this compound is quite soluble in water. For example, Wikipedia notes the solubility of Manganese(II) sulfate in water is 52 g/100 mL at 5 °C and 70 g/100 mL at 70 °C. Manganese(II) sulfate tetrahydrate should have similar solubility rates with correction for the hydration state.

      Please note that this information has not been validated internally:
      https://en.wikipedia.org/wiki/Manganese(II)_sulfate

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service