Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02425

SAFC

Chloral hydrate

EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP

Synonym(s):

Chloral hydrate, Trichloroacetaldehyde hydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
165.4
MDL number:
UNSPSC Code:
12352200
EC Index Number:
206-117-5

agency

BP
JP
Ph. Eur.
USP

Quality Level

vapor pressure

13 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

solid

potency

479 mg/kg LD50, oral (Rat)
3030 mg/kg LD50, skin (Rat)

pH

3.5-5.5 (20 °C, 100 g/L in H2O)

bp

97 °C/1013 hPa (decomposes)

mp

52 °C

solubility

6600 g/L

density

1.91 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C2H3Cl3O2/c3-2(4,5)1(6)7/h1,6-7H

InChI key

RNFNDJAIBTYOQL-UHFFFAOYSA-N

General description

EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP

Analysis Note

Assay (alkalimetric)99.5 - 101.0 %Identitypasses testAppearance of solution (Ph Eur)passes testAppearance of solution (JP)passes testAcidic substancespasses testpH-value (10 %; water)3.5 - 5.5Chloride (Cl)≤ 0.007 %Heavy metals (as Pb)≤ 0.0020 %Chloral alcoholatepasses testBenzenepasses testToluene≤ 890 ppmOther residual solvents (ICH Q3C)excluded by production processSubstances discolored by H₂SO₄passes testNon volatile matter≤ 0.1 %Sulfated ash≤ 0.10 %Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).Corresponds to Ph Eur, BP, JP

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_c

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Quality Management Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service