Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERD-121

Supelco

Dimethyl hydrogen phosphate solution

1000 μg/mL in methanol, ampule of 1.2 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H7O4P
CAS Number:
Molecular Weight:
126.05
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

1000 μg/mL in methanol

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

COP([O-])(=O)OC

InChI

1S/C2H7O4P/c1-5-7(3,4)6-2/h1-2H3,(H,3,4)/p-1

InChI key

KKUKTXOBAWVSHC-UHFFFAOYSA-M

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zoubeida Dhaouadi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 73(5), 805-814 (2009-05-16)
The effect of hexahydrated monovalent and divalent cations on the geometrical and vibrational features of dimethyl phosphate, dimethyl phosphorothioate and dimethyl phosphorodithioate anions (simple suitable model compounds representing the anionic moieties of natural and some modified nucleic acids) was studied.
Bhanu P Jena
Methods in cell biology, 90, 183-198 (2009-02-07)
Target membrane proteins, SNAP-25 and syntaxin (t-SNARE), and secretory vesicle-associated membrane protein (v-SNARE), are part of the conserved protein complex involved in fusion of opposing bilayers in biological systems in the presence of calcium. It is known that SNARE interaction
N D Forsberg et al.
Xenobiotica; the fate of foreign compounds in biological systems, 41(5), 422-429 (2011-03-31)
Levels of urinary dialkylphosphates (DAPs) are currently used as a biomarker of human exposure to organophosphorus insecticides (OPs). It is known that OPs degrade on food commodities to DAPs at levels that approach or exceed those of the parent OP.
Maria G Margariti et al.
Environmental research, 109(7), 821-829 (2009-08-14)
Hair analysis for dialkyl phosphates' (DAPs) residues could provide a measure of chronic exposure to organophosphate pesticides (OPs). The aim of this study was to determine whether these metabolites can be internally incorporated into the hair of rabbits exposed to
Amitava Adhikary et al.
The journal of physical chemistry. B, 116(20), 5900-5906 (2012-05-05)
Neutral sugar radicals formed in DNA sugar-phosphate backbone are well-established as precursors of biologically important damage such as DNA strand scission and cross-linking. In this work, we present electron spin resonance (ESR) evidence showing that the sugar radical at C5'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service