Key Documents
Z741824
Extech Temperature, Humidity Datalogger
About This Item
UNSPSC Code:
41100000
NACRES:
NB.61
feature
CE compliant
resolution 0.1 %RH
resolution 0.1 °F/C
(WEEE Compliant)
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments 42270
parameter
-40-185 °F temperature (-40-85 °C)
size
3 in. × 2.3 in. × 0.88 in., 76 x 57.2 x 22.3mm
weight
2.6 oz72.6g
General description
Compact, powerful and easy-to-use dataloggers record up to 16,000 readings. Multiple Dataloggers can be programmed and data downloaded from one single docking station. USB cable not included.
Choose from one of the most recent versions:
Certificates of Analysis (COA)