Key Documents
Z564303
Buchi® Rotavapor® distillation chiller, B-741
AC/DC input 230 V AC
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
feature
CE compliant
CSA approved
manufacturer/tradename
Büchi 046770
temperature display measuring range
10 °C
temperature measuring range
15-32 °C
power consumption
500 W (at 15°C)
General description
The ideal replacement for cooling water for the Rotavapor. Has a compact design and castors to provide space-saving set-up.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)