Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z407658

Büchi® Model B-580 oven, with drying tube assembly

AC/DC input 115 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

manufacturer/tradename

Brinkman 15-15-000-9

Looking for similar products? Visit Product Comparison Guide

General description

The B-580 glass oven is suitable for the drying, distillation, or sublimation of 100 to 250 mL samples under inert gas or vacuum if required. A glass tube with an electrically conducting coating acts as the heating element. The sample is heated in an appropriate drying tube or flask and can be viewed through the glass oven walls to monitor the process.
Specifications:
  • Digital temperature display
  • Temp. range: 40 to 300 °C
  • Temp. accuracy: ± 5 °C (in center of oven at 300 °C)
  • Warm-up time: from 20 to 300 °C in approx. 10 min
  • CE compliant
Drying tube assembly consists of tube which enters oven and end cap fitted with stopcock with vacuum and atmosphere/inert gas connections. Sample is placed in drying tube, which attaches to end cap with vacuum-tight flange. End cap is designed to hold desiccant to absorb water driven off by heat. Oven may be used for sublimation using optional sublimation insert.

Legal Information

Büchi is a registered trademark of Büchi Labortechnik AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service