Key Documents
Z407496
Sigma-Aldrich
Aldrich® swivel McLeod vacuum gauge
McLeod gauge (Mercury swivel)
About This Item
UNSPSC Code:
41112406
NACRES:
NB.79
description
Square root scale: 2μm to 5mm
linear scale 0 - 5 mm in 0.1mm div.
General description
Made with permanent linear and square root ceramic scales on a white background, high-vacuum O-ring seals, precision bore capillaries, Delrin® plastic and black anodized aluminum parts. Accurate to ±3% of reading or 1mm of scale division, whichever is greater. Gauges include operating instructions but do not include oil or mercury fill. Swivel arrangement permits rigid connection to the vacuum system with the scales vertical at all times.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
recommended
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)