Key Documents
Z261866
Cylinder valve and cap wrench
pkg of 1 ea
About This Item
UNSPSC Code:
41100000
packaging
pkg of 1 ea
manufacturer/tradename
Matheson TW-5
General description
Enables easy opening of gas cylinder valves that have been fitted with a hand wheel. It is also the correct and safe tool for removing cylinder caps.
Choose from one of the most recent versions:
Certificates of Analysis (COA)