Key Documents
Z243167
Prentice Hall molecular model set, 2/E student set, organic chemistry
this kit enables users to build virtually all simple molecules encountered in organic chemistry
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
feature
no. of atoms 60 Molymod centers
no. of bonds 92 bonding links
manufacturer/tradename
Pearson Education
General description
This kit enables users to build virtually all simple molecules encountered in organic chemistry. Includes space-filling models that simulate the true shape of saturated compounds. Provides open models that form realistic single, double, and triple bonds - even strained rings. Allows smooth rotation of the bonds to make conformational analysis easy. Contains enough components to create several models at once.
The components are precision-tooled from quality plastics, virtually indestructible and comes in a sturdy plastic case for easy storage. Provides a useful Instruction Book — with photos, diagrams, and concise discussions of chemical principles.
Set contains:
60 Molymod atom centers (7 types) and 92 bonding links (40 single-medium length, 12 double-long length, and 40 single or space-filling short bonds).
Scale is 2.8 cm = 100 pm (open style) and 1.45 cm = 100 pm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)