Key Documents
Z198668
Hazardous Materials NFPA - Classification sign
W × L 10 in. × 14 in.
About This Item
UNSPSC Code:
41100000
packaging
pkg of 10
W × L
10 in. × 14 in.
General description
Post in areas where NFPA Hazard Rating Systems labels are used for quick reference. The Hazardous Materials NFPA-Classification sign graphically explains the NFPA 704 Hazard Rating System. 0.030in. rigid PS.
Choose from one of the most recent versions:
Certificates of Analysis (COA)