Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W779989

Sigma-Aldrich

L-Cysteine hydrochloride monohydrate

FG, produced by Wacker Chemie AG, Burghausen, Germany, Food, 20 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
Beilstein/REAXYS Number:
5158059
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Flavis number:
17.032
NACRES:
NA.21

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 178/2002

assay

98.5-101.0%

form

powder or crystals

optical activity

[α]/D 5.7 to 6.8° (Ph. Eur. (2.2.7))

impurities

≤10 ppm heavy metals (Ph. Eur. (2.4.8))
≤200 ppm ammonium (Ph. Eur. (2.4.1))

ign. residue

≤0.1% (Ph. Eur (2.4.14))

loss

8-12% loss on drying

particle size

20 mesh

solubility

water: 25 mg/mL, clear, colorless

cation traces

As: ≤1 ppm (Ph. Eur. (2.4.2))
Pb: ≤5 ppm

suitability

clear, colorless for appearance of solution (Ph. Eur. (2.2.1))
positive for identity (Ph. Eur. (2.2.24)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

odorless

SMILES string

O.Cl.N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m0../s1

InChI key

QIJRTFXNRTXDIP-JIZZDEOASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

NMDA glutamatergic receptor antagonist.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerdt Müller et al.
Biotechnology for biofuels, 8, 187-187 (2015-11-27)
The emerging bioeconomy depends on improved methods for processing of lignocellulosic biomass to fuels and chemicals. Saccharification of lignocellulose to fermentable sugars is a key step in this regard where enzymatic catalysis plays an important role and is a major
Qinjian Zhao et al.
Human vaccines & immunotherapeutics, 10(3), 734-739 (2013-12-05)
Cryo-transmission electron microscopy (cryoTEM) is a powerful characterization method for assessing the structural properties of biopharmaceutical nanoparticles, including Virus Like Particle-based vaccines. We demonstrate the method using the Human Papilloma Virus (HPV) VLPs in GARDASIL®. CryoTEM, coupled to automated data
Edward T Chouchani et al.
Nature medicine, 19(6), 753-759 (2013-05-28)
Oxidative damage from elevated production of reactive oxygen species (ROS) contributes to ischemia-reperfusion injury in myocardial infarction and stroke. The mechanism by which the increase in ROS occurs is not known, and it is unclear how this increase can be
Feng Wang et al.
Cell, 153(6), 1379-1393 (2013-06-12)
Some species mount a robust antibody response despite having limited genome-encoded combinatorial diversity potential. Cows are unusual in having exceptionally long CDR H3 loops and few V regions, but the mechanism for creating diversity is not understood. Deep sequencing reveals
Jiahua Li et al.
American journal of physiology. Cell physiology, 305(2), C190-C196 (2013-05-17)
Claudins constitute a family of tight junction transmembrane proteins whose first extracellular loop (ECL1) determines the paracellular permeability and ion selectivity in epithelia. There are two cysteines in the ECL1 that are conserved among all claudins. We hypothesized that these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service