Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W778893

Sigma-Aldrich

α-Cyclodextrin

FG, produced by Wacker Chemie AG, Burghausen, Germany, Food

Synonym(s):

β-CD, Cavamax® W6, α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
Beilstein/REAXYS Number:
79627
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

grade

FG

reg. compliance

EU Regulation 178/2002

description

derived from GMO corn

impurities

≤0.5% reducing substances (determined as dextrose)
≤20 ppm volatile organics
≤5 ppm heavy metals
0 Salmonella/E.Coli in 10 g

ign. residue

≤0.1%

loss

≤11% loss on drying

mp

>278 °C (dec.) (lit.)

cation traces

As: ≤1.3 ppm
Pb: ≤0.5 ppm

application(s)

flavors and fragrances

food allergen

no known allergens

organoleptic

odorless

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C36H60O30/c37-1-7-25-13(43)19(49)31(55-7)62-26-8(2-38)57-33(21(51)15(26)45)64-28-10(4-40)59-35(23(53)17(28)47)66-30-12(6-42)60-36(24(54)18(30)48)65-29-11(5-41)58-34(22(52)16(29)46)63-27-9(3-39)56-32(61-25)20(50)14(27)44/h7-54H,1-6H2/t7-,8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-/m1/s1

InChI key

HFHDHCJBZVLPGP-RWMJIURBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Cyclodextrin (α-CD) is a cyclic oligosaccharide that consists of six glucopyranose units linked by α-(1,4) bonds. It can form a stable complex with dietary fat, which is resistant to lipolytic hydrolysis and is excreted intact leading to weight loss.[1]

Application

α-CD can act as a stabilizer for emulsions such as mayonnaise, margarine or butter creams.[2] The addition of α-CD (5-10%) to soy protein hydrolysate solution led to a decrease in its bitterness due to the formation of inclusion complexes with its amino acids.[3]

Legal Information

Cavamax is a registered trademark of Wacker Chemie AG

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taste modification of amino acids and protein hydrolysate by a-cyclodextrin
Linde GA, et al.
Food Research International, 42(7), 814-818 (2009)
Cyclodextrins and their uses: a review
Del Valle EMM
Process. Biochem., 39(9), 1033-1046 (2004)
The Beneficial Effects a-Cyclodextrin on Blood Lipids and Weight Loss in Healthy Humans
Comerford KB, et al
Obesity (Silver Spring, Md.), 19(6), 1200-1204 (2011)
Michael Kundel et al.
Analytical chemistry, 84(3), 1439-1445 (2012-01-17)
Here we present a new application of a time-of-flight aerosol mass spectrometer (TOF-AMS) for the measurement of atmospheric trace gases in real-time. Usually, TOF-AMS instruments are not sensitive to gas-phase species due to the aerodynamic particle focusing inlet system which
Yuya Hayashi et al.
Amyloid : the international journal of experimental and clinical investigation : the official journal of the International Society of Amyloidosis, 19 Suppl 1, 47-49 (2012-04-24)
RNA interference (RNAi) is a sequence-specific gene-silencing mechanism triggered by double-stranded RNA and powerful tools for a gene function study and RNAi therapy. Although siRNAs offer several advantages as potential new drugs to treat various diseases, the efficient delivery system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service