Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W530358

Sigma-Aldrich

Chamomile absolute

natural, FG

Synonym(s):

Chamomile oil, blue, Chamomile oil, Hungarian, German chamomile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Matricaria chamomilla L.

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 182.20

refractive index

n20/D 1.502
n20/D 1.540

density

0.930 g/mL at 25 °C (lit.)
1.016 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

floral; rich; warm

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

199.4 °F

flash_point_c

93 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service