Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W530241

Sigma-Aldrich

Rice bran oil

Certified organic (NOP)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164502
grade:
Certified organic (NOP)
Kosher

grade

Certified organic (NOP)
Kosher

Quality Level

origin

Mexico origin

refractive index

n20/D 1.472

density

0.919 g/mL at 25 °C

application(s)

flavors and fragrances

Legal Information

Certified organic by MOSA

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>212.0 °F

flash_point_c

> 100 °C

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhihua Song et al.
Applied biochemistry and biotechnology, 168(2), 364-374 (2012-09-06)
The ability of immobilized lipase from Rhizomucor miehei (Lipozyme RM IM) to catalyze the reaction of high-acid rice bran oil (RBO) and monoglyceride (MG) for diacylglycerol-enriched rice bran oil (RBO-DG) preparation was investigated. The effects of substrate ratio, reaction temperature
Somsuvra B Ghatak et al.
Phytotherapy research : PTR, 26(11), 1701-1708 (2012-03-13)
Nutrition and nutritional status can influence the development, maintenance and optimal functioning of the immune system. Oryzanol (OZ), a commercially important bioactive phytochemical component of rice bran oil, has generated global interest on account of its several beneficial physiological effects
Akanksha Kanitkar et al.
Bioresource technology, 102(17), 7896-7902 (2011-07-01)
Transesterification of vegetable oils (from soybeans and rice bran) into methyl and ethyl esters using a batch microwave system was investigated in this study. A critical comparison between the two alcohols was performed in terms of yields, quality, and reaction
Hussein O Ammar et al.
International journal of pharmaceutical compounding, 16(6), 516-523 (2012-12-25)
Rice bran oil and its bioactive constituents have been reported to possess different health benefit effects. Rice bran oil-containing pharmaceutical and cosmeceutical formulae are dispensed and characterized to elucidate the impact of their properties on clinical applications. Liquisolid and semisolid
Ratchanee Charoen et al.
Journal of food science, 76(1), E165-E172 (2011-05-04)
Rice bran oil (RBO) is used in foods, cosmetics, and pharmaceuticals due to its desirable health, flavor, and functional attributes. We investigated the effects of biopolymer emulsifier type and environmental stresses on the stability of RBO emulsions. Oil-in-water emulsions (5%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service