Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W530119

Sigma-Aldrich

Almond oil, sweet

Certified organic (NOP)

Synonym(s):

Almond oil from Prunus dulcis, Almond oil, sweet

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502
organoleptic:
almond
grade:
Certified organic (NOP)
Fragrance grade
Halal
Kosher
agency:
follows IFRA guidelines
food allergen:
tree nuts

grade

Certified organic (NOP)
Fragrance grade
Halal
Kosher

Quality Level

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

origin

Kenya origin

refractive index

n20/D 1.471

density

0.910 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

tree nuts

fragrance allergen

no known allergens

organoleptic

almond

Looking for similar products? Visit Product Comparison Guide

General description

Sweet almond oil is generally obtained from the dried kernels of the almond plant. It is traditionally used as massage oil and also as a natural emollient.[1]

Legal Information

Certified organic by MOSA

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hossein Saidi et al.
Human & experimental toxicology, 31(5), 518-522 (2011-04-22)
Aluminum phosphide (ALP), as an effective pesticide and a substance used for protecting rice during storage, has become one of the commonest causes of poisoning and even suicide in developing countries including Iran and India. The authors aimed to study
Ammar Cherif et al.
Comptes rendus biologies, 332(12), 1069-1077 (2009-11-26)
The evolution of the composition of sterols and squalene during the maturation of the fruit of three cultivars (Achaak, Perlees and Mazetto) almond (Prunus amygdalus Batsh) was investigated. At the same time the evolution of oleic, linoleic and linolenic fatty
Gianpiera Spada et al.
Carbohydrate polymers, 92(1), 40-47 (2012-12-12)
Two water in oil emulsions composed by eudermic ingredients as glycerin, cocoa butter, almond oil and a variety of lipids, were enriched respectively with milk thistle dry extract (MT) or with a binary complex composed by MT and hydroxypropyl-β-cyclodextrin (HP)
Natália Mezzomo et al.
Bioresource technology, 101(14), 5622-5632 (2010-03-06)
Peach kernels are industrial residues from the peach processing, contain oil with important therapeutic properties and attractive nutritional aspects because of the high concentration of oleic and linoleic acids. The extraction method used to obtain natural compounds from raw matter
Sophie Gallier et al.
Food & function, 3(5), 547-555 (2012-02-23)
An aqueous suspension of almond oil bodies (about 10% lipids) was prepared and subjected to in vitro gastric (with pepsin) and intestinal (with bile salts and pancreatin) digestion, simulating fasting conditions. The physicochemical and structural changes of the almond oil

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service