Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W512850

Sigma-Aldrich

Lavender oil

synthetic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502

biological source

synthetic

optical activity

[α]25/D -10 to -3°, neat

refractive index

n20/D 1.459-1.470
n20/D 1.461 (lit.)

bp

204 °C (lit.)

density

0.875-0.888 g/mL at 25 °C
0.879 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

lavender; herbaceous; floral

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

168.8 °F - closed cup

flash_point_c

76 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siegfried Kasper et al.
Wiener medizinische Wochenschrift (1946), 160(21-22), 547-556 (2010-12-21)
We review the data on the efficacy and tolerability of silexan, a novel preparation from lavender oil for oral use, in the treatment of anxiety disorders and related condition with particular attention to subthreshold generalized anxiety disorder (GAD). Three randomized
Catriona Jones
Complementary therapies in clinical practice, 17(4), 215-220 (2011-10-11)
Anecdotal evidence suggests that some women find lavender oil effective at relieving some of the symptoms associated with perineal trauma following childbirth. This paper examines the literature detailing the efficacy of lavender oil in wound care in an attempt to
Tomoe Ueno-Iio et al.
Life sciences, 108(2), 109-115 (2014-06-10)
Lavender essential oil (Lvn) has been reported to have anti-inflammatory effects. Bronchial asthma is characterized by bronchial allergic inflammation with airway remodeling. Therefore, we evaluated the anti-inflammatory effect of Lvn on experimentally induced bronchial asthma in a murine model. BALB/c

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service